Få mere at vide
Thallium Standard For ICP/MS, SPEX CertiPrep™
Artikelnummer. 15246016 Shop alle SPEX Certiprep produkter
Mængde:
125 mL
Pakningsstørrelse:
125ml
Artikelnummer. 15246016
Brand: SPEX Certiprep CLTL21BY
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Tl1μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Thallium Standard
Navn note ICP/MS
Emballage Triple-leached LDPE bottle
CAS 7440-28-0, 7697-37-2, 7732-18-5
Koncentration eller sammensætningsnoter Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyt- eller komponentnavne Thallium Standard
Fysisk form Fluid
Molekylær formel Tl
MDL nummer MFCD00134063
Til brug med (udstyr) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI nøgle BKVIYDNLLOSFOA-UHFFFAOYSA-N
SMIL [Tl]
Element Th
IUPAC navn thallium
Opløsningsmiddel eller Matrix 2% HNO3
Molekylvægt (g/mol) 204.38
PubChem CID 5359464
Mængde 125 mL
ChEBI CHEBI:30440
Grad Claritas PPT
Koncentration eller sammensætning (efter analyt eller komponenter) 1mg/L
Vis mere Vis mindre
Product intended for laboratory use only