Få mere at vide
Thallium(I) Trifluoromethanesulfonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Thallium(I) Trifluoromethanesulfonate |
| CAS | 73491-36-8 |
| Molekylær formel | CF3O3STl |
| Mængde | 1 g |
| Synonym | Thallium Triflate, Thallium Trifluoromethanesulfonate, Thallium(1+) Triflate, Thallium(1+) Trifluoromethanesulfonate, Thallous Triflate, 1,1,1-Trifluoro-methanesulfonic Acid Thallium(1+) Salt |
| InChI formel | InChI=1S/CHF3O3S.Tl/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1 |
| SMIL | O=S(C(F)(F)F)(O[Tl])=O |
| IUPAC navn | thallium(I) trifluoromethanesulfonate |
| Molekylvægt (g/mol) | 353.45 |
| Formel vægt | 353.93 |