Få mere at vide
MedChemExpress Thalidomide-O-amido-PEG1-(C1-PEG)2-C2-NH2 TFA
Artikelnummer. 16415496
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
1 g
2 g
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16415496 100 mg 100mg
16481606 500 mg -
16461179 1 g -
16430146 2 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16415496 Leverandør MedChemExpress Leverandørnr. HY112600A
MedChemExpress Thalidomide-O-amido-PEG1-(C1-PEG)2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology.
Tekniske data
Molekylvægt (g/mol) 648.58 g/mol
CAS 2435572-49-7
Indhold og opbevaring 4°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Forsendelsestilstand Room temperature
Mængde 100 mg
Synonym Cereblon Ligand-Linker Conjugates 12 TFA ∣ E3 Ligase Ligand-Linker Conjugates 23 TFA