Få mere at vide
Thalidomide-O-amido-C6-NH2 TFA, MedChemExpress
Artikelnummer. 16481179
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
1 g
2 g
Pakningsstørrelse:
2g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16481179 1 g -
16475496 100 mg -
16450146 2 g 2g
16401616 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16481179 Leverandør Medchem Express Leverandørnr. HY112618A
MedChemExpress Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA), a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, can be used in the synthesis of PROTACs.
Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA), a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, can be used in the synthesis of PROTACs.
Tekniske data
Kemisk navn eller materiale Thalidomide-O-amido-C6-NH2 TFA
CAS 1950635-14-9
Farve White
Renhedsgrad noter Research
Molekylær formel C23H27F3N4O8
Mængde 1 g
Synonym Cereblon Ligand-Linker Conjugates 11 TFAE3 Ligase Ligand-Linker Conjugates 25 TFA
Opløselighedsinformation DMSO : ≥ 105.5 mg/mL (193.76 mM)
SMIL O=C(NCCCCCCN)COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O.O=C(O)C(F)(F)F
Molekylvægt (g/mol) 544.48
Formel vægt 544.48
Procent renhed 99.82%
Grad Research
Fysisk form Solid