Få mere at vide
Thalidomide - linker 2, Tocris Biosciences™
GREENER_CHOICE
PROTACs (Proteolysis Targeting Chimeras)
Brand:  Tocris Bioscience 6468/25
Encompass
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Produktkode 15927960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Thalidomide 4'-oxyacetamide-PEG4-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 2
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
PROTAC Toolbox
2245697-85-0
Thalidomide - linker 2
25 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner