Få mere at vide
Thalidomide-d4, TRC
Artikelnummer. 30451275
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451275 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451275 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3388521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thalidomide-d4
CAS 1219177-18-0
Molekylær formel C13 2H4 H6 N2 O4
Mængde 1 mg
Synonym 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4','2-(2,6-dioxo-3-piperidinyl)- (ACI), 2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione-4,5,6,7-d4 (ACI), 4,5,6,7-Tetradeuterio-2-(2,6-dioxopiperidin-3-yl)isoindole-1,3-dione, (±)-Thalidomide-d4 (phenyl-d4), Thalidomide-D4 (phenyl-D4), Thalidomide-d4
InChI formel InChI=1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2C(=O)N(C3CCC(=O)NC3=O)C(=O)c2c1[2H]
IUPAC navn 4,5,6,7-tetradeuterio-2-(2,6-dioxopiperidin-3-yl)isoindole-1,3-dione
Molekylvægt (g/mol) 262.25
Formel vægt 262.0892