Få mere at vide
Tocris Bioscience™ Thalidomide 4'-oxyacetamide-alkylC4-azide
Beskrivelse
Thalidomide 4'-oxyacetamide-alkylC4-azide is a click-activated Thalidomide (Cat.No. 0652); precursor to PROTAC® that hijacks cereblon as the E3 ubiquitin ligase component. Supplied with an azide functional handle at a position known not to significantly affect binding to cereblon, for ready click conjugation to a linker/target protein ligand.
This product has been recently renamed. The previous name for this product was Azido-Thalidomide
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| CAS | 2098488-36-7 |
| Synonym | Azido-Thalidomide |
| Mængde | 50 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C19H20N6O6 |
| Formel vægt | 428.4 |
| Renhed | 98% |