Få mere at vide
Tocris Bioscience™ Thalidomide 4'-ether-alkylC6-acid
Beskrivelse
Thalidomide 4'-ether-alkylC6-acid is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC6 linker with a terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 7-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]heptanoic acid |
| CAS | 2169266-69-5 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C20H22N2O7 |
| Renhed | 0.95 |