Få mere at vide
TH 588, Tocris Bioscience™
Potent MTH1 inhibitor
Brand:  Tocris Bioscience 5334/25
Encompass
Produktkode 15943221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
TH 588 is a potent MTH1 inhibitor (IC50 = 5 nM). Induces oxidative DNA damage and reduces survival in cancer cells. Inhibits growth of breast and colorectal cancer xenografts in mice.
Tekniske data
Tekniske data
N4-Cyclopropyl-6-(2,3-dichlorophenyl)-2,4-pyrimidinediamine
1609960-31-7
>98%
25 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner