Få mere at vide
TH 588, Tocris Bioscience™
Potent MTH1 inhibitor
Brand: Tocris Bioscience 5334/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TH 588 is a potent MTH1 inhibitor (IC50 = 5 nM). Induces oxidative DNA damage and reduces survival in cancer cells. Inhibits growth of breast and colorectal cancer xenografts in mice.Tekniske data
| N4-Cyclopropyl-6-(2,3-dichlorophenyl)-2,4-pyrimidinediamine | |
| 1609960-31-7 | |
| >98% | |
| 25 mg |