missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ TGDS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112739
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TGDS Polyclonal Antibody for Western Blot, IHC (P), ELISA
Tekniske data
| TGDS | |
| Polyclonal | |
| Unconjugated | |
| TGDS | |
| 2610017J16Rik; 2610025M23Rik; AI648925; CATMANS; dTDP-D-glucose 4,6-dehydratase; growth-inhibiting protein 21; SDR2E1; short chain dehydrogenase/reductase family 2E, member 1; TDPGD; TDP-glucose 4,6-dehydratase; TGDS | |
| Rabbit | |
| Protein G | |
| RUO | |
| 23483, 306164 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O95455 | |
| TGDS | |
| Recombinant Human dTDP-D-glucose 4,6-dehydratase protein (143-305AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion