missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies TG101348

Artikelnummer. 17120232
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17120232 10 mg
17110232 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17120232 Leverandør STEMCELL Technologies Leverandørnr. 73474

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

JAK/STAT pathway inhibitor; Inhibits JAK2

  • TG101348 is an inhibitor of Janus kinase 2 (JAK2) with an IC50 of 6 nM
  • It also inhibits related kinases fms-related tyrosine kinase 3 (FLT3), RET, and JAK3 with less potent activity, having IC50 values of 25, 17 and 169 nM, respectively (Pardanani et al.) It is proposed to bind to the ATP binding pocket of its kinase targets (Zhou et al.)
  • Inhibits growth of Ba/F3 cells expressing JAK2 V617F or MPL W515L mutations (Pardanani et al.)
  • Reduces tumor cell burden and increases survival in mouse models of JAK2 V617F-induced hematopoietic and myeloproliferative disease (Pardanani et al.; Wernig et al.)
  • Sensitizes erlotinib-resistant non-small cell lung cancer cells to erlotinib treatment in vitro and in a mouse xenograft model (Zhang et al.)
  • Displaces BRD4 from chromatin and suppresses c-MYC expression in multiple myeloma cells in vitro (Ciceri et al.)

Tekniske data

CAS 936091-26-8
Synonym Fedratinib; SAR302503
Mængde 10 mg
Molekylær formel C27H36N6O3S
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.