Learn More
Ebro Electronic™ TFX Core Thermometer Series
Precise and very robust thermometer with a big display
Brand: Ebro Electronic™ 1340-5417A
This item is not returnable.
View return policy
Description
Thermometer for various applications in food, laboratory, industial use. All thermometers includes a factory calibration certificate. The thermometer measures with PT1000 temperature sensor.
Specifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
For bakeries, butcheries and for the food industry | |
Calibration Cert., No Cable | |
120 mm | |
TFX 410-1 and TPX 100 | |
0.1°C | |
Precision Core Thermometer | |
Without cable | |
300°C | |
-50°C | |
Yes | |
Approx. 5 Years | |
±0.3°C/±0.5°F | |
LCD | |
IP67 |
Single-use | |
52 mm | |
335 mm | |
Stainless Steel, Plastic | |
Blunt | |
0.3 kg | |
None | |
572°F | |
-58°F | |
-50°C to +300°C | |
AG 130, AG 140, AG 150, AG 160, AX 100 | |
3.0 V Lithium Battery Exchangeable | |
120 mm | |
50 mm |
Product Content Correction
Product Title
Ebro Electronic™ TFX Core Thermometer Series > with immersion probe
Spot an opportunity for improvement?Share a Content Correction