Learn More
Cytiva (formerly Pall Lab) TF PTFE Membrane Disc Filters
Strong, chemically resistant membranes for air monitoring and sampling in aggressive environments.
Brand:  Cytiva (formerly Pall Lab) 66149
Additional Details : Weight : 0.05100kg
 View more versions of this product
Product Code. 17194541
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Low chemical background permits highly sensitive, interference-free determinations
  • Ensures accurate gravimetric determinations with low tare mass
  • Ideal for filtration of gas and/or organic solvents
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PTFE
0.45 μm
Non-sterile
47 mm