missing translation for 'onlineSavingsMsg'
Learn More

Tetrapentylammonium bromide, 99+%, Thermo Scientific Chemicals

Catalog Number p-3779651
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
866-97-7
C20H44BrN
378.48
MFCD00011856
SPALIFXDWQTXKS-UHFFFAOYSA-M
tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
70086
[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
This item is not returnable. View return policy
866-97-7
C20H44BrN
378.48
MFCD00011856
SPALIFXDWQTXKS-UHFFFAOYSA-M
tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
70086
[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 102°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)4]4NBr
Quantity 5 g
Beilstein 04, III, 331
IUPAC Name tetrapentylazanium bromide
Formula Weight 378.47
Percent Purity 99+%
Physical Form Shiny Flakes
Chemical Name or Material Tetrapentylammonium bromide
Show More Show Less
compliance-icons
Product Identifier
  • Tetrapentylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'