missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TETRAN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2695.00 DKK - 3850.00 DKK
Specifications
Antigen | TETRAN |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18126867
|
Bio-Techne
NBP2-47607 |
0.1 mL |
3850.00 DKK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18622526
|
Novus Biologicals
NBP2-47607-25ul |
25 ÎĽL |
2695.00 DKK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TETRAN Polyclonal specifically detects TETRAN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TETRAN | |
Polyclonal | |
Rabbit | |
Human | |
major facilitator superfamily domain containing 10, major facilitator superfamily domain-containing protein 10 | |
MFSD10 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10227 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FLPETLPLEKRAPSIALGFRDAADLLS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TETRAN Antibody, Novus Biologicals™