Få mere at vide
Tetramisole Hydrochloride, Mikromol™
Beskrivelse
Tetramisole Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tetramisole hydrochloride |
| CAS | 5086-74-8 |
| Analyt- eller komponentnavne | Tetramisole hydrochloride |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C11 H12 N2 S . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Tetramisole Hydrochloride, Imidazo[2,1-b]thiazole, 2,3,5,6-tetrahydro-6-phenyl-, hydrochloride (1:1), Imidazo[2,1-b]thiazole, 2,3,5,6-tetrahydro-6-phenyl-, monohydrochloride (9CI), Imidazo[2,1-b]thiazole, 2,3,5,6-tetrahydro-6-phenyl-, monohydrochloride, (+/-)- (8CI), (+/-)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, (+/-)-Tetramisole hydrochloride, 2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Anthelvet, Citarin, Concurat, Curaminth, DL-Tetramisole hydrochloride, McN JR 8299, NSC 170985, NSC 215179, Nilverm, Nilverom, R 8299, RP 16535, Ripercol, Tetraamisol hydrochloride, Verminject |
| InChI formel | InChI=1S/C11H12N2S.ClH/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10;/h1-5,10H,6-8H2;1H |
| SMIL | Cl.C1CN2CC(N=C2S1)c3ccccc3 |
| IUPAC navn | 6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride |
| Vis mere |