Få mere at vide
Tetramisole hydrochloride, MedChemExpress
Beskrivelse
Tetramisole hydrochloride is an inhibitor of alkaline phosphatases, is a high purity antiparasitic.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tetramisole hydrochloride |
| Fysisk form | Solid |
| CAS | 5086-74-8 |
| Farve | White |
| Mængde | 2 g |
| Renhedsgrad noter | Research |
| Molekylær formel | C11H13ClN2S |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | (±)-Tetramisole hydrochloride DL-Tetramisole hydrochloride R-829 |
| Opløselighedsinformation | H2O : 100 mg/mL (415.37 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (41.54 mM; Need ultrasonic) |
| Vis mere |