Learn More
Tetramethylsilane-d12, TRC
Product Code. 30423182
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30423182 5 mg 5mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30423182 Supplier Toronto Research Chemicals Supplier No. TRCT3029095MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetramethylsilane-D12
CAS 18145-38-5
Molecular Formula C4D12Si
Quantity 5 mg
Synonym Tetra(methyl-d3)silane, Tetrakis(trideuteromethyl)silane, Tetramethylsilane-d12
InChI Formula InChI=1S/C4H12Si/c1-5(2,3)4/h1-4H3/i1D3,2D3,3D3,4D3
SMILES [2H]C([2H])([2H])[Si](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name tetrakis(methyl-d3)silane
Molecular Weight (g/mol) 100.3
Formula Weight 100.15