Få mere at vide
Tetramethyl-d12-urea, CDN
Artikelnummer. 30322686
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322686 0.05 g
30310361 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322686 Leverandør CDN Leverandørnr. CDND05740.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetramethylurea-D12
CAS 51219-89-7
Molekylær formel C5 D12 N2 O
Mængde 0.05 g
Synonym Urea, tetra(methyl-d3)- (9 CI), 1,1,3,3-Tetrakis(trideuteriomethyl)urea, Tetramethylurea-d12, Tetra(methyl-d3)urea, Tetramethylurea-D12
InChI formel InChI=1 S/C5H12N2O/c1-6(2)5(8)7(3)4/h1-4H3/i1D3,2D3,3D3,4D3
SMIL [2 H]C([2 H])([2 H])N(C(=O)N(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,3,3-tetrakis(trideuteriomethyl)urea
Molekylvægt (g/mol) 128.236
Formel vægt 128.17 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity