Få mere at vide
Tetramethyl Bisphenol A-d6, TRC
Artikelnummer. 30415077
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30415077 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415077 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3009321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetramethyl Bisphenol A-d6
CAS 203578-30-7
Molekylær formel C19 D6 H18 O2
Mængde 1 mg
Synonym 2,2-Bis[3,5-(dimethyl-d6)-4-hydroxyphenyl]propane, Bisxylenol A-d6, NSC 73730-d6, 2,2-Bis[4-hydroxy-3,5-(dimethyl-d6)phenyl]propane, Tetramethylbisphenol A-d6, 3,3',5,5'-Tetramethyl-4,4'-dihydroxydiphenyl-2,2-propane-d6, 4,4'-Isopropylidenebis[2,6-(dimethyl-d6)phenol], 4,4'-Isopropylidenedi-2,6-xylenol-d6
InChI formel InChI=1S/C19H24O2/c1-11-7-15(8-12(2)17(11)20)19(5,6)16-9-13(3)18(21)14(4)10-16/h7-10,20-21H,1-6H3/i5D3,6D3
SMIL [2H]C([2H])([2H])C(c1cc(C)c(O)c(C)c1)(c2cc(C)c(O)c(C)c2)C([2H])([2H])[2H]
IUPAC navn 4-[1,1,1,3,3,3-hexadeuterio-2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]-2,6-dimethylphenol
Molekylvægt (g/mol) 290.43
Formel vægt 290.215