missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetrakis(triphenylphosphine)nickel(0)

Brand:  Alfa Aesar™ 032101.03

Additional Details : CAS Number : 15133-82-1 Weight : 0.00100kg Transport : UN number : 1325 Chem class : 4.1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15471247

  • 962.08 DKK / 1g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

15133-82-1
1107.86
KFBKRCXOTTUAFS-UHFFFAOYSA-N
11979963
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ni]
C72H60NiP4
MFCD00010011
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
nickel;triphenylphosphane

Specifications

Tetrakis(triphenylphosphine)nickel(0)
Brown
1g
C72H60NiP4
MFCD00010011
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
KFBKRCXOTTUAFS-UHFFFAOYSA-N
nickel;triphenylphosphane
11979963
Crystalline
Air and moisture sensitive
123°C to 128°C
15133-82-1
Ni[P(C6H5)3]4
UN1325
Insoluble in water.
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ni]
1107.86
1107.83
Videos
SDS Documents