missing translation for 'onlineSavingsMsg'
Få mere at vide

Tetrakis(acetonitril)palladium(II) Bis(trifluoromethanesulfonat) >90%, TRC

Product Code. 30427303
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30427303 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427303 Supplier Toronto Research Chemicals Supplier No. TRCT29633525MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tetrakis(acetonitrile)palladium(II) Bis(trifluoromethanesulfonate)
CAS 68569-14-2
Molekylær formel C8 H12 N4 Pd . C F3 O3 S
Mængde 25 mg
Synonym Tetrakis(acetonitrile)palladium Bis(triflate), Tetrakis(acetonitrile)palladium Ditriflate, Tetrakis(acetonitrile)palladium(2+) Ditriflate, (SP-4-1)-Tetrakis(acetonitrile)palladium(2+) Salt With Trifluoromethanesulfonic Acid
InChI formel InChI=1S/4C2H3N.CHF3O3S.Pd/c4*1-2-3;2-1(3,4)8(5,6)7;/h4*1H3;(H,5,6,7);/q;;;;;+2/p-1
SMIL CC#[N][Pd+2]([N]#CC)([N]#CC)[N]#CC.[O-]S(=O)(=O)C(F)(F)F
IUPAC navn tetrakis(ethylidyne-5-azanyl λ)palladium(2+); trifluoromethanesulfonat
Molekylvægt (g/mol) 419.697
Formel vægt 418.961
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.