missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydroxyquinone, Honeywell Fluka™

Catalog Number p-7120869
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5676-48-2
C6H8O8
208.122
MFCD00001597
PTIMJXRLFTZAOV-UHFFFAOYSA-N
19849282
2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione;dihydrate
C1(=C(C(=O)C(=C(C1=O)O)O)O)O.O.O
This item is not returnable. View return policy
5676-48-2
C6H8O8
208.122
MFCD00001597
PTIMJXRLFTZAOV-UHFFFAOYSA-N
19849282
2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione;dihydrate
C1(=C(C(=O)C(=C(C1=O)O)O)O)O.O.O

Titration Indicator (for Ba, SO4)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
UN Number NONH for all modes of transport
Beilstein 1371548
Formula Weight 208.12g/mol
Packaging Glass bottle
Quantity 1 g
Physical Form Powder
Chemical Name or Material Tetrahydroxyquinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'