Få mere at vide
Tetrahydropyrimidine-2(1H)-thione, TRC
Artikelnummer. 30454509
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454509 100 mg
30453187 500 mg
30452253 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30454509 Leverandør Toronto Research Chemicals Leverandørnr. TRCT311371100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2055-46-1
Molekylær formel C4H8N2S
Molekylvægt (g/mol) 116.18
Synonym Tetrahydro-2(1H)-pyrimidinethione, 1,3-Diazinane-2-thione, 1,4,5,6-Tetrahydro-2-pyrimidinethiol, 2-Mercapto-1,4,5,6-tetrahydropyrimidine, 2-Mercapto-3,4,5,6-tetrahydropyrimidine, 2-Mercaptotetrahydropyrimidine, 3,4,5,6-Tetrahydro-1H-pyrimidine-2-thione, 3,4,5,6-Tetrahydro-2(1H)-pyrimidinethione, 3,4,5,6-Tetrahydro-2-mercaptopyrimidine, 3,4,5,6-Tetrahydro-2-pyrimidinethiol, Cyclic propylene thiourea, Hexahydropyrimidine-2-thione, N,N'-Trimethylenethiourea, NSC 21316, Perhydropyrimidine-2-thione, Tetrahydropyrimidine-2-thione, Trimethylenethiourea
IUPAC navn 1,3-diazinane-2-thione
SMIL S=C1NCCCN1
Tekniske data
Kemisk navn eller materiale Tetrahydropyrimidine-2(1H)-thione
Mængde 100 mg
InChI formel InChI=1S/C4H8N2S/c7-4-5-2-1-3-6-4/h1-3H2,(H2,5,6,7)
Formel vægt 116.0408