Få mere at vide
Tetrahydroharmine, Mikromol™
Artikelnummer. 17927764
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17927764 100 mg
17907964 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17927764 Leverandør Mikromol Leverandørnr. MM2562.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tetrahydroharmine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tetrahydro-Harmine
CAS 17019-01-1
Analyt- eller komponentnavne tetrahydro-Harmine
Forsendelsestilstand Room Temperature
Molekylær formel C13 H16 N2 O
Certificeringer/compliance ISO 9001
Synonym 1H-Pyrido[3,4-b]indole, 2,3,4,9-tetrahydro-7-methoxy-1-methyl-, Harmine, tetrahydro- (6CI), 1,2,3,4-Tetrahydroharmine, Leptaflorine, Tetrahydroharmine, 1,2-Dihydroharmaline, (+/-)-Tetrahydroharmine
InChI formel InChI=1S/C13H16N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-4,7-8,14-15H,5-6H2,1-2H3
SMIL COc1ccc2c3CCNC(C)c3[nH]c2c1
IUPAC navn 7-methoxy-1-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
Molekylvægt (g/mol) 216.28
Mængde 100 mg
Formel vægt 216.1263 g/mol