Få mere at vide
Tetrahydrodehydrocorticosterone, TRC
Artikelnummer. 30134585
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134585 250 mg
30011532 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134585 Leverandør Toronto Research Chemicals Leverandørnr. TRCT293165250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 566-03-0
Molekylær formel C21 H32 O4
Molekylvægt (g/mol) 348.48
Synonym Pregnane-11,20-dione, 3,21-dihydroxy-, (3α,5β)-, 5β-Pregnane-11,20-dione, 3α,21-dihydroxy- (8CI), (3α,5β)-3,21-Dihydroxypregnane-11,20-dione, 11-Dehydrotetrahydrocorticosterone, 3α,21-Dihydroxy-5β-pregnane-11,20-dione, 5β-Pregnane-3α,21-diol-11,20-dione, NSC 11574, Tetrahydro Kendall's compound A, Tetrahydro-11-dehydrocorticosterone, Tetrahydrodehydrocorticosterone
IUPAC navn (3R,5R,8S,9S,10S,13S,14S,17S)-3-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,3,4,5,6,7,8,9,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-11-one
SMIL C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)CC(=O)[C@H]23
Tekniske data
Kemisk navn eller materiale Tetrahydrodehydrocorticosterone
Mængde 250 mg
InChI formel InChI=1S/C21H32O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12-16,19,22-23H,3-11H2,1-2H3/t12-,13-,14+,15+,16-,19-,20+,21+/m1/s1
Formel vægt 348.2301