Få mere at vide
Tetrahydroabietyl Alcohol, TRC
Artikelnummer. 30179531
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179531 50 mg
30179159 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179531 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29304550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13393-93-6
Molekylær formel C20 H36 O
Molekylvægt (g/mol) 292.5
Synonym 1-Phenanthrenemethanol, tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-, Abietyl alcohol, tetrahydro- (6CI), (1R,4aR,4bS,10aR)-Tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenemethanol, Tetrahydroabietyl alcohol, Tetrahydroabietyl Alcohol
IUPAC navn (1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,8,8a,9,10,10a-dodecahydrophenanthren-1-yl)methanol
SMIL CC(C)C1CCC2C(CCC3C(C)(CO)CCCC23C)C1
Tekniske data
Kemisk navn eller materiale Tetrahydroabietyl Alcohol
Mængde 50 mg
InChI formel InChI=1S/C20H36O/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4/h14-18,21H,5-13H2,1-4H3
Formel vægt 292.2766