Få mere at vide
Tetrahydroabietic Acid (Stereoisomer), TRC
Artikelnummer. 30426574
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426574 10 mg
30423832 25 mg
30416773 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426574 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29447010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 358976-89-3
Molekylær formel C20H34O2
Molekylvægt (g/mol) 306.48
Synonym Tetrahydro-abietic Acid, (1R,4aR,4bS,10aR)-Tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic Acid, (1R,4aR,4bS,10aR)-7-Isopropyl-1,4a-dimethyltetradecahydrophenanthrene-1-carboxylic Acid
IUPAC navn (1R,4aR,4bS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,8,8a,9,10,10a-dodecahydrophenanthrene-1-carboxylic acid
SMIL C[C@@]12[C@](CCC3[C@]2([H])CCC(C(C)C)C3)([H])[C@@](C(O)=O)(C)CCC1
Tekniske data
Kemisk navn eller materiale Tetrahydroabietic Acid (Stereoisomer)
Mængde 10 mg
InChI formel InChI=1S/C20H34O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h13-17H,5-12H2,1-4H3,(H,21,22)/t14?,15?,16-,17+,19+,20+/m0/s1
Formel vægt 306.2559