Få mere at vide
Tetrahydro Cortisone-d5, TRC
Artikelnummer. 30423741
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30423741 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423741 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2933780.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetrahydro Cortisone-d5
CAS 1485501-48-1
Molekylær formel C21 D5 H27 O5
Mængde 0.5 mg
Synonym (3α,5β)-3,17,21-Trihydroxypregnane-11,20-dione-d5, 3α,17,21-Trihydroxy-5β-pregnan-11,20-dione-d4, 5β-Pregnan-3α,17α,21-triol- 11,20-dione-d5, 5β-Tetrahydrocortisone-d5, Ba 2681-d5, NSC 76984-d5, THE-d5, Tetrahydro E-d5, Tetrahydrocompound E-d5, Urocortisone-d5, 5beta-Pregnan-3alpha,17alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5beta-Pregnan-3alpha,17alpha,21-triol-11,20-dione-9,12,12,21,21-d5
InChI formel InChI=1S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,18,22-23,26H,3-11H2,1-2H3/t12-,13-,14+,15+,18-,19+,20+,21+/m1/s1/i10D2,11D2,18D
SMIL [2H]C([2H])(O)C(=O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@@]3([2H])C(=O)C([2H])([2H])[C@]12C
IUPAC navn (3R,5R,8S,9S,10S,13S,14S,17R)-9,12,12-trideuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-3,17-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-11-one
Molekylvægt (g/mol) 369.51
Formel vægt 369.2564