missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-3830072
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16874-33-2
C5H8O3
116.12
MFCD00022473
UJJLJRQIPMGXEZ-UHFFFAOYSA-N
2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
86079
oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O
This item is not returnable. View return policy
16874-33-2
C5H8O3
116.12
MFCD00022473
UJJLJRQIPMGXEZ-UHFFFAOYSA-N
2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
86079
oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
Color Yellow to Brown
CAS Max % 100.0
Density 1.1700g/mL
Boiling Point 128.0°C to 129.0°C (13.0 mmHg)
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4590 to 1.4610
Assay Percent Range 98% min. (GC)
Specific Gravity 1.17
Solubility Information Solubility in water: soluble
Formula Weight 116.12
Percent Purity 98+%
Chemical Name or Material Tetrahydro-2-furoic acid, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'