Få mere at vide
Tetrahydro-2-[(2R)-1,1,2-trimethyl-3-(phenylsulfonyl)propoxy]-2H-pyran-d6, TRC
Artikelnummer. 30415786
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30415786 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415786 Leverandør Toronto Research Chemicals Leverandørnr. TRCT295822100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetrahydro-2-[(2R)-1,1,2-trimethyl-3-(phenylsulfonyl)propoxy]-2H-pyran-d6
CAS 1796539-93-9
Molekylær formel C17H20D6O4S
Mængde 100 mg
InChI formel InChI=1S/C17H26O4S/c1-14(13-22(18,19)15-9-5-4-6-10-15)17(2,3)21-16-11-7-8-12-20-16/h4-6,9-10,14,16H,7-8,11-13H2,1-3H3/t14-,16?/m0/s1/i2D3,3D3
SMIL [2H]C([2H])([2H])C(OC1CCCCO1)([C@@H](C)CS(=O)(=O)c2ccccc2)C([2H])([2H])[2H]
IUPAC navn 2-[(3R)-4-(benzenesulfonyl)-1,1,1-trideuterio-3-methyl-2-(trideuteriomethyl)butan-2-yl]oxyoxane
Molekylvægt (g/mol) 332.49
Formel vægt 332.1928