Få mere at vide
Tetraethylenepentamine Pentahydrochloride, TRC
Artikelnummer. 30426038
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426038 10 g
30418310 5 g
30418309 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426038 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29151510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4961-41-5
Molekylær formel C8H28Cl5N5
Molekylvægt (g/mol) 371.61
Synonym N1-(2-Aminoethyl)-N2-[2-[(2-aminoethyl)amino]ethyl]-1,2-ethanediamine Hydrochloride (1:5)
IUPAC navn N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine;pentahydrochloride
SMIL Cl.Cl.Cl.Cl.Cl.NCCNCCNCCNCCN
Tekniske data
Kemisk navn eller materiale Tetraethylenepentamine Pentahydrochloride
Mængde 10 g
InChI formel InChI=1S/C8H23N5.5ClH/c9-1-3-11-5-7-13-8-6-12-4-2-10;;;;;/h11-13H,1-10H2;5*1H
Formel vægt 369.0787