Få mere at vide
Tetraethyleneglycol Bisibuprofen Ester, TRC
Artikelnummer. 30006227
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30006227 250 mg -
30005387 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006227 Leverandør Toronto Research Chemicals Leverandørnr. TRCT292280250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1161014-75-0
Molekylær formel C34 H50 O7
Molekylvægt (g/mol) 570.757
Synonym Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, 1,1'-[oxybis(2,1-ethanediyloxy-2,1-ethanediyl)] ester, 2-[2-[2-[2-[2-(4-isobutylphenyl)propanoyloxy]ethoxy]ethoxy]ethoxy]ethyl 2-(4-isobutylphenyl)propanoate, 1,1'-[Oxybis(2,1-ethanediyloxy-2,1-ethanediyl)
IUPAC navn 2-[2-[2-[2-[2-[4-(2-methylpropyl)phenyl]propanoyloxy]ethoxy]ethoxy]ethoxy]ethyl 2-[4-(2-methylpropyl)phenyl]propanoate
SMIL CC(C)Cc1ccc(cc1)C(C)C(=O)OCCOCCOCCOCCOC(=O)C(C)c2ccc(CC(C)C)cc2
Tekniske data
Kemisk navn eller materiale 1,1'-[Oxybis(2,1-ethanediyloxy-2,1-ethanediyl)] 2-(4-Isobutylphenyl)propanoate
Mængde 250 mg
InChI formel InChI=1S/C34H50O7/c1-25(2)23-29-7-11-31(12-8-29)27(5)33(35)40-21-19-38-17-15-37-16-18-39-20-22-41-34(36)28(6)32-13-9-30(10-14-32)24-26(3)4/h7-14,25-28H,15-24H2,1-6H3
Formel vægt 570.356