missing translation for 'onlineSavingsMsg'
Få mere at vide

Tetraethyldithio Pyrophosphate/Sulfotepp, SPEX CertiPrep™

Artikelnummer. 12959374 Shop alle SPEX Certiprep produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mL
Pakningsstørrelse:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
12959374 1 mL 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 12959374 Leverandør SPEX Certiprep Leverandørnr. S3455

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Indeholder

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Tetraethyldithio Pyrophosphate/Sulfotepp3689-24-51000μg/mL

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Kemisk navn eller materiale Tetraethyldithio Pyrophosphate/Sulfotepp
Emballage 2mL amber prescored glass ampule
CAS 3689-24-5, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Metoder For US EPA Methods 622
Analyt- eller komponentnavne Tetraethyldithio Pyrophosphate/Sulfotepp
Molekylær formel C8H20O5P2S2
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle XIUROWKZWPIAIB-UHFFFAOYSA-N
SMIL CCOP(=S)(OCC)OP(=S)(OCC)OCC
Element Tetraethyldithio Pyrophosphate, Sulfotepp
IUPAC navn diethoxyphosphinothioyloxy-diethoxy-sulfanylidene-$l^{5}-phosphane
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 322.311
PubChem CID 19395
Mængde 1 mL
ChEBI CHEBI:38945
Vis mere Vis mindre

Product intended for laboratory use only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.