Få mere at vide
Tetraethylammonium Cyanide, TRC
Artikelnummer. 30425353
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30425353 1 g
1 options
Artikelnummer. 30425353 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2927151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetraethylammonium Cyanide
CAS 13435-20-6
Molekylær formel C8H20N.CN
Mængde 1 g
Synonym N,N,N-Triethylethanaminium Cyanide, Tetraethylammonium Cyanide, N,N,N-triethylethanaminium Hydrocyanic Acid
InChI formel InChI=1S/C8H20N.CN/c1-5-9(6-2,7-3)8-4;1-2/h5-8H2,1-4H3;/q+1;-1
SMIL CC[N+](CC)(CC)CC.[C-]#N
IUPAC navn tetraethylazanium;cyanide
Molekylvægt (g/mol) 156.27
Formel vægt 156.1626