Få mere at vide
Tetraethyl Pyrophosphite, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tetraethyl Pyrophosphite |
| CAS | 21646-99-1 |
| Molekylær formel | C8H20O5P2 |
| Mængde | 1 g |
| Synonym | Ethyl Pyrophosphite, Pyrophosphorous Acid','Tetraethyl Ester, Tetraethyl Diphosphite, Tetraethyl Pyrophosphite, diethoxyphosphanyl diethyl phosphite, EINECS 244-494-8 |
| InChI formel | InChI=1S/C8H20O5P2/c1-5-9-14(10-6-2)13-15(11-7-3)12-8-4/h5-8H2,1-4H3 |
| SMIL | O(P(OCC)OCC)P(OCC)OCC |
| IUPAC navn | diethoxyphosphanyl diethyl phosphite |
| Molekylvægt (g/mol) | 258.19 |
| Formel vægt | 258.08 |