Få mere at vide
Tetradehydropodophyllotoxin, TRC
Artikelnummer. 30415101
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415101 1 g
30412401 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415101 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2914451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42123-27-3
Molekylær formel C22H18O8
Molekylvægt (g/mol) 410.37
Synonym 9-Hydroxy-5-(3,4,5-trimethoxyphenyl)-furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(8H)-one, Dehydropodophyllotoxin
IUPAC navn 9-hydroxy-5-(3,4,5-trimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
SMIL O=C(OC1)C2=C1C(O)=C(C=C(OCO3)C3=C4)C4=C2C5=CC(OC)=C(OC)C(OC)=C5
Tekniske data
Kemisk navn eller materiale Tetradehydropodophyllotoxin
Mængde 1 g
InChI formel InChI=1S/C22H18O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,23H,8-9H2,1-3H3
Formel vægt 410.1