Få mere at vide
Tetradecyl diphenylphosphate, TRC
Artikelnummer. 30426804
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426804 1 g
30423188 100 mg
30412770 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426804 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2926531G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142474-86-0
Molekylær formel C26H39O4P
Molekylvægt (g/mol) 446.56
IUPAC navn diphenyl tetradecyl phosphate
SMIL CCCCCCCCCCCCCCOP(=O)(Oc1ccccc1)Oc2ccccc2
Tekniske data
Kemisk navn eller materiale Tetradecyl diphenylphosphate
Mængde 1 g
InChI formel InChI=1S/C26H39O4P/c1-2-3-4-5-6-7-8-9-10-11-12-19-24-28-31(27,29-25-20-15-13-16-21-25)30-26-22-17-14-18-23-26/h13-18,20-23H,2-12,19,24H2,1H3
Formel vægt 446.2586