Få mere at vide
Tetradecyl Acetate, TRC
Artikelnummer. 30418345
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30418345 50 mg
1 options
Artikelnummer. 30418345 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29170050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetradecanol acetate
CAS 638-59-5
Molekylær formel C16 H32 O2
Mængde 50 mg
Synonym 1-Tetradecanol','acetate (7CI,8CI,9CI), 1-Tetradecyl acetate, Acetic acid tetradecan-1-yl ester, Myristyl acetate, Tetradecanol acetate, Tetradecyl acetate, n-Tetradecyl acetate
InChI formel InChI=1S/C16H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h3-15H2,1-2H3
SMIL CCCCCCCCCCCCCCOC(=O)C
IUPAC navn tetradecyl acetate
Molekylvægt (g/mol) 256.42
Formel vægt 256.2402