Få mere at vide
Tetradecanoic-14,14,14-d3 Acid, CDN
Artikelnummer. 30318907
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318907 0.1 g 0.1g
30321211 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318907 Leverandør CDN Leverandørnr. CDND36040.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetradecanoic-14,14,14-D3 Acid
CAS 62217-71-4
Molekylær formel C14 D3 H25 O2
Mængde 0.1 g
Synonym Tetradecanoic-14,14,14-d3 acid (9 CI), Myristic-14,14,14-d3 acid (7 CI), 14,14,14-Trideuteriotetradecanoic acid, [14,14,14-2H3]Tetradecanoic acid, Tetradecanoic acid-14,14,14-d3
InChI formel InChI=1 S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCC(=O)O
IUPAC navn 14,14,14-trideuteriotetradecanoic acid
Molekylvægt (g/mol) 231.3894
Formel vægt 231.2278 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity