Få mere at vide
Tetradecanoic-12-d1 Acid, CDN
Artikelnummer. 30325589
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325589 0.1 g
30325354 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325589 Leverandør CDN Leverandørnr. CDND55110.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetradecanoic-12-D Acid
CAS 358730-97-9
Molekylær formel C14 D H27 O2
Mængde 0.1 g
Synonym Tetradecanoic-12-d acid (9 CI), Myristic acid-12-d, Tetradecanoic acid-12-d, Tetradecanoic-12-D acid
InChI formel InChI=1 S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i3D
SMIL [2 H]C(CC)CCCCCCCCCCC(=O)O
IUPAC navn 12-deuteriotetradecanoic acid
Molekylvægt (g/mol) 229.377
Formel vægt 229.215 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity