Learn More
WTW™ TetraCon™ DU/T Flow-through Conductivity Cell
Flow measuring cells by WTW for medium and high conductivity are suitable for extensive measuring series in the lab but also for the maintenance and measurement of cleaning solutions in special systems.
Brand: WTW™ 301252
6885.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Specifications
0.778 cm-1 | |
Epoxy, Graphite | |
TetraCon™ DU/T |
Flow-through Conductivity Cell | |
4-electrode, temperature sensor |