Learn More
WTW TetraCon 925/C IDS Conductivity probe, 0.475 cm
Brand:  WTW 301721
6677.56 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
TetraCon 925/C IDS Conductivity probe, 0.475 cm
Product Code. 17522785
  • 6850.00 DKK / Each
Explore more special offers
Product Title
WTW TetraCon 925/C IDS Conductivity probe, 0.475 cm >