Få mere at vide
Tetracaine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tetracaine hydrochloride |
| CAS | 136-47-0 |
| Analyt- eller komponentnavne | Tetracaine hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H24 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Tetracaine Hydrochloride, Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Ethanol, 2-dimethylamino-, p-butylaminobenzoate, HCl (4CI), 2-(Dimethylamino)ethyl p-(butylamino)benzoate hydrochloride, 2-Dimethylaminoethanol 4-n-butylaminobenzoate hydrochloride, Amethocaine hydrochloride, Amethocaine monohydrochloride, Anacel, Anethain, Anethaine, Butethanol, Curtacain, Decicain, Decicaine, Dessicain, Dianguang, Dicaine, Dikain, Intercaine, Leocaine, Menonasal, Niphanoid, Pantocain, Pantocaine, Pantocaine hydrochloride, Pantocaine monohydrochloride, Pontocaine hydrochloride, Tetocaine, Tetracaine chlorhydrate, Tetracaine hydrochloride, Tetracaine monohydrochloride, Tetracainium chloride, Tonexol, p-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester hydrochloride |
| InChI formel | InChI=1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C |
| Vis mere |