Få mere at vide
Tetrabutylphosphonium 2H,2H-Perfluorodecanoate, TRC
Artikelnummer. 30135073
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135073 100 mg
30135261 50 mg
17982325 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135073 Leverandør Toronto Research Chemicals Leverandørnr. TRCT310655100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 882489-14-7
Molekylær formel C16 H36 P . C10 H2 F17 O2
Molekylvægt (g/mol) 736.53
Synonym Phosphonium, tetrabutyl-, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoate (1:1), Phosphonium, tetrabutyl-, salt with 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoic acid (1:1) (9CI), Tetrabutylphosphonium 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecanoate, 8:2 FTCA tetrabutylphosphonium salt, 8:2 Fluorotelomer carboxylic acid tetrabutylphosphonium salt, Perfluoro-octylethanoic acid tetrabutylphosphonium salt, Tetrabutylphosphonium 2H,2H-perfluorodecanoate
IUPAC navn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoate;tetrabutylphosphanium
SMIL CCCC[P+](CCCC)(CCCC)CCCC.[O-]C(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Tetrabutylphosphonium 2H,2H-Perfluorodecanoate
Mængde 100 mg
InChI formel InChI=1S/C16H36P.C10H3F17O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;11-3(12,1-2(28)29)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h5-16H2,1-4H3;1H2,(H,28,29)/q+1;/p-1
Formel vægt 736.2338