Learn More
Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Catalog Number p-195860
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
429-42-5
C16H36BF4N
329.27
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
429-42-5
C16H36BF4N
329.27
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Beilstein 04, III, 293
Formula Weight 329.28
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium tetrafluoroborate, 95%
compliance-icons
Product Identifier
  • Tetrabutylammonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.