Learn More
Tetrabutylammonium Difluorotriphenylsilicate, TRC
Product Code. 30412481
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412481 25 g 25g
30411286 5 g -
30420578 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30412481 Supplier Toronto Research Chemicals Supplier No. TRCT29124025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 163931-61-1
Molecular Formula C18 H15 F2 Si . C16 H36 N
Molecular Weight (g/mol) 539.858
Synonym N,N,N-Tributyl-1-butanaminium (TB-5-11)-Difluorotriphenylsilicate(1-), TBAT, Tetrabutylammonium Triphenyldifluorosilicate, Tetrabutylammonium Triphenylsilyl Difluoride, Tetrabutylammonium(1+) Difluorotriphenylsilicate(1-)
IUPAC Name difluoro(triphenyl)silanuide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.F[Si-](F)(c1ccccc1)(c2ccccc2)c3ccccc3
Specifications
Quantity 25 g
InChI Formula InChI=1S/C18H15F2Si.C16H36N/c19-21(20,16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h1-15H;5-16H2,1-4H3/q-1;+1
Formula Weight 539.376
Chemical Name or Material Tetrabutylammonium Difluorotriphenylsilicate
Product Title
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.