missing translation for 'onlineSavingsMsg'
Få mere at vide

Tetrabutylammonium bibenzoate, TRC

Artikelnummer. 30413144
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413144 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30413144 Leverandør Toronto Research Chemicals Leverandørnr. TRCT291230250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Tetrabutylammonium bibenzoate
CAS 116263-39-9
Molekylær formel C30H47NO4
Mængde 250 mg
Synonym N,N,N-Tributyl-1-butanaminium Bibenzoate, Bioxyanion
InChI formel InChI=1S/C16H36N.2C7H6O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2*8-7(9)6-4-2-1-3-5-6/h5-16H2,1-4H3;2*1-5H,(H,8,9)/q+1;;/p-1
SMIL CCCC[N+](CCCC)(CCCC)CCCC.OC(=O)c1ccccc1.[O-]C(=O)c2ccccc2
IUPAC navn benzoic acid;tetrabutylazanium;benzoate
Molekylvægt (g/mol) 485.7
Formel vægt 485.3505
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.