Få mere at vide
Tetrabenzylamino-tetramethyl Cyclen p-TSA salt, TRC
Artikelnummer. 30450910
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450910 50 mg
30450892 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30450910 Leverandør Toronto Research Chemicals Leverandørnr. TRCT75505050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetrabenzylamino-tetramethyl Cyclen p-TSA salt
CAS 488106-79-2
Molekylær formel C40H52N4 x C7H8O3S
Mængde 50 mg
Synonym (2S,5S,8S,11S)-4-Methylbenzenesulfonate-2,5,8,11-tetramethyl-1,4,7,10-tetrakis(phenylmethyl)-1,4,7,10-tetraazacyclododecane
InChI formel InChI=1S/C40H52N4.C7H8O3S/c1-33-25-42(30-38-19-11-6-12-20-38)35(3)27-44(32-40-23-15-8-16-24-40)36(4)28-43(31-39-21-13-7-14-22-39)34(2)26-41(33)29-37-17-9-5-10-18-37;1-6-2-4-7(5-3-6)11(8,9)10/h5-24,33-36H,25-32H2,1-4H3;2-5H,1H3,(H,8,9,10)/t33-,34-,35-,36-;/m0./s1
SMIL C[C@H]1CN(Cc2ccccc2)[C@@H](C)CN(Cc3ccccc3)[C@@H](C)CN(Cc4ccccc4)[C@@H](C)CN1Cc5ccccc5.Cc6ccc(cc6)S(=O)(=O)O
IUPAC navn 4-methylbenzenesulfonic acid;(2S,5S,8S,11S)-1,4,7,10-tetrabenzyl-2,5,8,11-tetramethyl-1,4,7,10-tetrazacyclododecane
Molekylvægt (g/mol) 761.08
Formel vægt 760.4386