Få mere at vide
Tetra-n-butyl-d36-tin, CDN
Artikelnummer. 30316863
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316863 0.25 g -
30313190 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316863 Leverandør CDN Leverandørnr. CDND55630.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetrabutylstannane-d36
CAS 358731-92-7
Molekylær formel C16 2H36 Sn
Mængde 0.25 g
InChI formel InChI=1 S/4C4H9.Sn/c4*1-3-4-2;/h4*1,3-4H2,2H3;/i4*1D2,2D3,3D2,4D2;
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[Sn](C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn tetrakis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)stannane
Molekylvægt (g/mol) 383.39
Formel vægt 384.4099 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity